Human Recombinant TIGIT (from HEK293 Cells)
Catalog # 103815-460
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:100 µg
- Tag sequence:Fc Tag (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) at 5 μg/mL (100 μL/well) can bind Human TIGIT, Fc Tag (Cat. No. TIT-H5254) with a linear range of 0.6-5 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.313-10 μg/mL (Routinely tested). Serial dilutions of Human TIGIT Neutralizing antibody were added into Human TIGIT, Fc Tag (Cat. No. TIT-H5254): Biotinylated Human CD155, Fc,Avitag (Cat. No. CD5-H82F6) binding reactions. The half maximal inhibitory concentration (IC50) is 0.55834 μg/mL (Routinely tested). Loaded Human TIGIT, Fc Tag (Cat. No. TIT-H5254) on Protein A Biosensor, can bind Human CD155, His Tag (Cat. No. CD5-H5223) with an affinity constant of 0.98 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS assay shows that recombinant Human TIGIT, Fc Tag (Cat. No. TIT-H5254) can bind to 293T cell overexpressing human CD155. The concentration of TIGIT used is 0.1 μg/mL (Routinely tested).
- Gene ID:Q495A1-1
- Protein Synonyms:VSTM3|TIGIT|VSIG9
- Protein/Peptide Name:TIGIT
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:39.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 10 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103815-460
Specifications
About this item
Human TIGIT Protein, Fc Tag (MALS verified), ACROBiosystems