Human Recombinant SLC1A5 (from E. coli)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human SLC1A5 Protein Tag, TrxA Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q15758-1
- Protein Synonyms:RDRC|RD114|ASCT2M7VS1|AAAT|RDR|R16|RDRCFLJ31068|ATB(0)|ASCT2|M7V1ATBO|M7V1
- Protein/Peptide Name:SLC1A5
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:22.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together