Order Entry
United States
ContactUsLinkComponent
Human Recombinant SIRP alpha (from HEK293 Cells), Biotin
Human Recombinant SIRP alpha (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems
Certificates
Human Recombinant SIRP alpha (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human CD47, Fc Tag (Cat. No. CD7-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (Cat. No. SIA-H82E0) with a linear range of 0.078-0.625 μg/mL (QC tested).
  • Protein Synonyms:
    MYD1|PTPNS1|CD172A|SHPS1|P84|SIRPA|BIT|MFR
  • Protein/Peptide Name:
    SIRP alpha
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    39.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together