Human Recombinant Siglec-9 (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human Siglec-9 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human Siglec-9, His Tag (Cat. No. SI9-H52H4) at 10 μg/mL (100 μL/well) on Nickel Coated plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-2.5 μg/mL (QC tested).
- Gene ID:Q9Y336-1
- Protein Synonyms:SIGLEC9|Siglec-9|CDw329|CD329
- Protein/Peptide Name:Siglec-9
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:37.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 25 mM MES, 150 mM NaCl, pH5.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together