Human Recombinant Siglec-8 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human Siglec-8 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q9NYZ4-1
- Protein Synonyms:SIGLEC8|SAF2|SAF-2|Siglec-8
- Protein/Peptide Name:Siglec-8
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:39.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS with Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together