Human Recombinant Siglec-15 (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human Siglec-15 / CD33L3 Protein, Mouse IgG2a Fc Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Gene ID:Q6ZMC9-1
- Protein Synonyms:Siglec-15|SIGLEC-15|Siglec15|CD33L3|CD33 antigen-like 3|sialic acid-binding Ig-like lectin 15
- Protein/Peptide Name:Siglec-15
- Molecular Weight:53.4 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together