Human Recombinant Serum Albumin (from HEK293 Cells), Biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human Serum Albumin Protein, His,Avitag™, ACROBiosystems
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Loaded Biotinylated Human Serum Albumin, His,Avitag (Cat. No. HSA-H82E3) on SA Biosensor, can bind Human FCGRT&B2M Heterodimer Protein, His Tag&Strep II Tag (SPR & BLI verified) (Cat. No. FCM-H5286) with an affinity constant of 0.641 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P02768-1
- Protein Synonyms:Serum albumin|Alb|ALB
- Protein/Peptide Name:Serum Albumin
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:70.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together