Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant PSCA (from HEK293 Cells)
 
Human Recombinant PSCA (from HEK293 Cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    O43653-1
  • Protein Synonyms:
    UNQ206|PRO232|PSCA
  • Protein/Peptide Name:
    PSCA
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    10.4 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

 

Frequently Bought Together