Human Recombinant PLAU (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human PLAU / uPA Protein, His Tag (activated by trypsin) (active enzyme), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P00749-1
- Protein Synonyms:URK|ATF|u-PA|Urokinase|PLAU|QPD|UPA|BDPLT5
- Protein/Peptide Name:PLAU
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:15.3 kDa, 17.8 kDa and 29.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 25 mM HEPES, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together