Human Recombinant PD-L1 (from HEK293 Cells), Biotin
Biotinylated Human PD-L1 / B7-H1 (19-134) Protein, His,Avitag™ (recommended for biopanning), ACROBiosystems
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human PD-L1, His,Avitag (Cat. No. PDL-H82E4) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) with a linear range of 0.8-13 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human PD-L1, His,Avitag (Cat. No. PDL-H82E4) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Anti-PD-L1 MAb, Human IgG1 with a linear range of 0.2-6 ng/mL (Routinely tested).
- Gene ID:Q9NZQ7-1
- Protein Synonyms:B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
- Protein/Peptide Name:PD-L1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:16.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together