Order Entry
United States
ContactUsLinkComponent
Human Recombinant PD-1 (from HEK293 Cells), Biotin
Human Recombinant PD-1 (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems
Certificates
Human Recombinant PD-1 (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    Fc, Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Loaded Biotinylated Human PD-1, Fc,Avitag,His Tag (Cat. No. PD1-H82F4) on SA Biosensor, can bind Human PD-L1, His Tag (HPLC verified) (Cat. No. PD1-H5229) with an affinity constant of 1.6 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Biotinylated Human PD-1, Fc,Avitag,His Tag (Cat. No. PD1-H82F4) on SA Biosensor, can bind PD-L2, His Tag (SPR verified) (Cat. No. PD2-H5220) with an affinity constant of 0.42 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q15116-1
  • Protein Synonyms:
    CD279|PDCD1|PD1|SLEB2
  • Protein/Peptide Name:
    PD-1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    45.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together