Human Recombinant OX40 Ligand (from HEK293 Cells), Biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human OX40 Ligand / TNFSF4 Protein, His,Avitag™, ACROBiosystems
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:His, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human OX40, Fc Tag (Cat. No. OX0-H5255) at 1 μg/mL (100 μL/well) can bind Biotinylated Human OX40 Ligand, His,Avitag (Cat. No. OXL-H82Q6) with a linear range of 0.4-6 ng/mL (QC tested).
- Gene ID:P23510-1
- Protein Synonyms:TNFSF4|CD134L|TXGP1|OX40L|Glycoprotein Gp34|CD134 ligand|CD252
- Protein/Peptide Name:OX40 Ligand
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:19.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together