Human Recombinant NKp46 (from HEK293 Cells), Biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human NKp46 / NCR1 / CD335 Protein, Fc,Avitag™, ACROBiosystems
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Precision Avi
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:AAH64806
- Protein Synonyms:CD335|NCR1|hNKp46|NK-p46|LY94
- Protein/Peptide Name:NKp46
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:54.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together