Human Recombinant Neuroligin-4
Catalog # 103790-696
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q8NFZ3-1
- Protein Synonyms:Y-linked|Neuroligin-4
- Protein/Peptide Name:Neuroligin-4, Y-linked
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:73.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-696
Specifications
About this item
Human Neuroligin-4, Y-linked / NLGN4Y Protein, His Tag, ACROBiosystems