Human Recombinant Nectin-4 (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human Nectin-4 Protein, Fc Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Loaded Human Nectin-4, Fc Tag (Cat. No. NE4-H5255) on Protein A Biosensor, can bind Human Nectin-1, His Tag (Cat. No. PV1-H5223) with an affinity constant of 0.263 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human Nectin-1, His Tag (Cat. No. PV1-H5223) on HIS1K Biosensor, can bind Human Nectin-4, Fc Tag (Cat. No. NE4-H5255) with an affinity constant of 2 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q96NY8-1
- Protein Synonyms:PVRL4|NECTIN4| PRR4|LNIR
- Protein/Peptide Name:Nectin-4
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:60.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together