Order Entry
United States
ContactUsLinkComponent
Human Recombinant Mucin-1 (from HEK293 Cells)
Human Recombinant Mucin-1 (from HEK293 Cells)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant Mucin-1 (from HEK293 Cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P15941-1
  • Protein Synonyms:
    Mucin 1|Episialin|CA 15-3|PEMT|PUM|MUC-1/X|CD227|MUC-1/SEC|KL-6|EMA|MUC1|MAM6|PEM|H23AG|MUC1/ZD
  • Protein/Peptide Name:
    Mucin-1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    60.2 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together