Human Recombinant Mesothelin (from HEK293 Cells), RFP Fusion Protein
Supplier: ACROBIOSYSTEMS
Certificates
About this item
RFP Fusion Human Mesothelin / MSLN (296-580) Protein, His Tag, ACROBiosystems
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:RFP Fusion Protein
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:RFP Fusion Human Mesothelin / MSLN (296-580) Protein, His Tag
- Storage Conditions:–20 ℃, aviod light
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Anti-Human MSLN Mab, Human IgG1 at 10 μg/mL (100 μL/well) can bind RFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HR2P4) with a linear range of 0.078-0.625 μg/mL (QC tested). can bind Human MS4A1 / CD20 Full Length Protein, His Tag (Cat. No. CD0-H55Ha)with an affinity constant of 1.21 nM as determined in a SPR assay (Biacore T200) (Routinely tested). 2e5 of the MSLN-CAR-293 cells were stained with 100 μL of 3 μg/mL of RFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HR2P4). Non-transfected 293 cells were used as negative control (QC tested).
- Protein Synonyms:MPF|MSLN|Mesothelin
- Protein/Peptide Name:Mesothelin
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:62.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together