Human Recombinant LIGHT (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human LIGHT / TNFSF14 Protein, Fc Tag (active trimer) (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (active trimer) (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Human LIGHT, Fc Tag (active trimer) (MALS verified) (Cat. No. LIT-H5269) induced cytotoxicity in HT-29 cells. The ED50 for this effect is 1.29-2.38 ng/mL (Routinely tested).
- Gene ID:O43557-1
- Protein Synonyms:HVEM-L|CD258|LTg|TR2|LIGHT|HVEML|CD258 antigen|TNFSF14
- Protein/Peptide Name:Light
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:82.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together