Human Recombinant LAG-3 (from HEK293 cells), PE (Phycoerythrin)
Catalog # 103815-222
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:PE (Phycoerythrin)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 tests
- Storage Conditions:–20 ℃, aviod light
- Gene ID:P18627-1
- Protein Synonyms:FDC|CD223|LAG3
- Protein/Peptide Name:LAG-3
- Molecular Weight:49.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS with Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103815-222
Specifications
About this item
PE-Labeled Human LAG-3 / CD223 Protein, His Tag (recommended for neutralizing assay), ACROBiosystems