Human Recombinant JAK1 (from Insect cells)
Human JAK1 Protein, His Tag, ACROBiosystems
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Storage Conditions:–70 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P23458-1
- Protein Synonyms:Jak1|JAK1B|JAK1|JAK-1|JAK1A
- Protein/Peptide Name:JAK1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:35.7 kDa
- Endotoxin Level:Low
- Formulation:Delivered as bulk protein in a 0.2 μm filtered solution of 50 mM Tris, 500 mM NaCl, 2 mM TCEP, pH8.0 with glycerol as protectant.
- Shipping Temperature:Dry Ice
Frequently Bought Together