Human Recombinant Integrin alpha E beta 7 (from HEK293 Cells)
Catalog # 103790-624
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P38570-1 (ITGAE) & P26010-1 (ITGB7)
- Protein Synonyms:Integrin alpha E beta 7|ITGAE&ITGB7
- Protein/Peptide Name:Integrin alpha E beta 7
- Purity:>90% as determined by reduced SDS-PAGE.
- Molecular Weight:128.6 kDa (ITGAE) & 81.8 kDa (ITGB7)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-624
Specifications
About this item
Human Integrin alpha E beta 7 (ITGAE&ITGB7) Heterodimer Protein, His Tag&Tag Free, ACROBiosystems