Human Recombinant Integrin alpha 10 beta 1 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human Integrin alpha 10 beta 1 (ITGA10&ITGB1) Heterodimer Protein, His Tag&Tag Free, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag&Tag Free
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Human ITGA10&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-H52Wa) with a linear range of 0.02-0.313 μg/mL (QC tested).
- Gene ID:O75578-1 (ITGA10) & P05556-1 (ITGB1)
- Protein Synonyms:Integrin alpha 10 beta 1|ITGA10&ITGB1
- Protein/Peptide Name:Integrin alpha 10 beta 1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:126.6 kDa (ITGA10) & 83.7 kDa (ITGB1)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together