Order Entry
United States
ContactUsLinkComponent
Human Recombinant Integrin alpha 1 beta 1 (from HEK293 Cells)
Human Recombinant Integrin alpha 1 beta 1 (from HEK293 Cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant Integrin alpha 1 beta 1 (from HEK293 Cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag&Tag Free
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Human ITGA1&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-H52Wb) with a linear range of 1-20 ng/mL (QC tested).
  • Gene ID:
    P56199-1 (ITGA1) & P05556-1 (ITGB1)
  • Protein Synonyms:
    ITGA1&ITGB1|Integrin alpha 1 beta 1
  • Protein/Peptide Name:
    Integrin alpha 1 beta 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    130.0 kDa (ITGA1) & 83.7 kDa (ITGB1)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together