Human Recombinant ILDR2 (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human ILDR2 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Storage Conditions:–20 °C
- Gene ID:Q71H61-1
- Protein Synonyms:C1orf32|ILDR2
- Protein/Peptide Name:ILDR2
- Molecular Weight:20.5 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together