Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL-23R (from HEK293 Cells)
Human Recombinant IL-23R (from HEK293 Cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant IL-23R (from HEK293 Cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc, Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human IL-23 R, Fc,Avitag (Cat. No. ILR-H82F3) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-H52W5) with a linear range of 39-156 ng/mL (QC tested). Measured by its inhibitory ability in a competitive ELISA. Immobilized Biotinylated Human IL-23 R, Fc,Avitag (Cat. No. ILR-H82F3) at 5 μg/mL (100 μL/well), can bind pre-mixed increasing concentrations of Human IL23A/p19 Neutralizing Antibody and 0.25 μg/mL (100 μL/well) Human IL-23A&IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-H52W5) with a half maximal inhibitory concentration (IC50) of 0.3889 μg/mL (Routinely tested).
  • Gene ID:
    Q5VWK5-1
  • Protein Synonyms:
    IL-23 R|IL-23 Receptor
  • Protein/Peptide Name:
    IL-23R
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    66.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together