Human Recombinant IL-2 R beta & IL-2 R gamma (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human IL-2 R beta&IL-2 R gamma Heterodimer Protein, Fc Tag&Fc Tag (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P14784-1 (IL-2RB) & P31785-1 (IL-2RG)
- Protein Synonyms:IL-2RB & IL-2RG|IL-2 R beta & IL-2 R gamma
- Protein/Peptide Name:IL-2 R beta & IL-2 R gamma
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:51.1 kDa (IL-2RB) and 53.6 kDa (IL-2RG)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together