Human Recombinant IL-2 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human IL-2 Protein, Fc,Avitag™, ACROBiosystems
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:Fc, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human IL-2, Fc,Avitag (Cat. No. IL2-H82F3) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Human IL-2 R alpha, His Tag (Cat. No. ILA-H52H9) with a linear range of 1-39 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Human IL-2 R alpha, Fc Tag (Cat. No. ILA-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-2, Fc,Avitag (Cat. No. IL2-H82F3) with a linear range of 0.6-10 ng/mL (Routinely tested). Loaded Biotinylated Human IL-2, Fc,Avitag (Cat. No. IL2-H82F3) on SA Biosensor, can bind Human IL-2 R beta, His Tag (SPR verified) (Cat. No. CD2-H5221) with an affinity constant of 2.4 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Biotinylated Human IL-2, Fc,Avitag (Cat. No. IL2-H82F3) on SA Biosensor, can bind Human IL-2 R alpha, His Tag (Cat. No. ILA-H52H9) with an affinity constant of 47.1 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P60568-1
- Protein Synonyms:TCGF|lymphokine|IL2|Interleukin 2
- Protein/Peptide Name:IL-2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:43.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together