Human Recombinant IL-17C (from HEK293 cells), Biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human IL-17C Protein, His,Avitag™, ACROBiosystems
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:His, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-17C, His,Avitag (Cat. No. ILC-H82E5) with a linear range of 0.5-13 ng/mL (QC tested).
- Gene ID:Q9P0M4-1
- Protein Synonyms:IL17C|Cytokine CX2|IL-17C
- Protein/Peptide Name:IL-17C
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:23.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together