Human Recombinant IL-17 RE (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human IL-17 RE (155-454) Protein, Fc Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human IL-17C, His Tag (Cat. No. ILC-H52H7) at 2 μg/mL (100 μL/well) can bind Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) with a linear range of 1-10 ng/mL (QC tested). Loaded Human IL-17 RE (155-454), Fc Tag (Cat. No. ILE-H5256) on Protein A Biosensor, can bind Human IL-17C, His Tag (Cat. No. ILC-H52H7) with an affinity constant of 0.714 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q8NFR9-1
- Protein Synonyms:IL-17 RE|IL-17RE|IL17RE|IL-17 receptor E
- Protein/Peptide Name:IL-17 RE
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:60.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together