Order Entry
United States
ContactUsLinkComponent
Human Recombinant Her2 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Human Recombinant Her2 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant Her2 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 ℃, aviod light
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Trastuzumab at 0.5 μg/mL (100 μL/well) can bind FITC-Labeled Human Her2, His Tag (Cat. No. HE2-HF224) with a linear range of 0.02-0.313 μg/mL (QC tested). 2e5 of Anti-Her2 CAR-293 were stained with 100 μL of 1 μg/mL of FITC-Labeled Human Her2, His Tag (Cat. No. HE2-HF224) and negative control protein respectively, FITC signal was used to evaluate the binding activity (QC tested).
  • Gene ID:
    P04626-1
  • Protein Synonyms:
    NEU|MLN19|TKR1|ERBB2|CD340|HER2|HER-2/neu|NGL
  • Protein/Peptide Name:
    Her2
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    70.2 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together