Order Entry
United States
ContactUsLinkComponent
Human Recombinant GUCY2C (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Human Recombinant GUCY2C (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant GUCY2C (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 ℃, aviod light
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    2e5 of Anti-GUCY2C CAR-293 cells were stained with 100 μL of 1 μg/mL of FITC-Labeled Human GUCY2C, His Tag (Cat. No. GUC-HF2H8) and negative control protein respectively, FITC signal was used to evaluate the binding activity(QC tested).
  • Gene ID:
    P25092-1
  • Protein Synonyms:
    GUCY2C|hSTAR|STA receptor|GC-C|STAR|GUC2C
  • Protein/Peptide Name:
    GUCY2C
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    47.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together