Human Recombinant Glypican 3 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human Glypican 3 / GPC3 Protein, Llama IgG2b Fc Tag, low endotoxin, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Llama IgG2b Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.01 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 μg/mL (100 μL/well) can bind Human Glypican 3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. GP3-H5257) with a linear range of 0.6-39 ng/mL (QC tested).
- Gene ID:P51654-1
- Protein Synonyms:GPC3|SGBS|SGB|Glypican-3|SDYS |MXR7|SGBS1|OCI5|GTR2-2|DGSX
- Protein/Peptide Name:Glypican 3
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:38.1 kDa and 50.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together