Human Recombinant GITR Ligand (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human GITR Ligand / TNFSF18 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human GITR Ligand, His Tag (Cat. No. GIL-H5249) at 2 μg/mL (100 μL/well) can bind Human GITR, Fc Tag (Cat. No. GIR-H5254) with a linear range of 0.1-4 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Human GITR Ligand, His Tag (Cat. No. GIL-H5249) at 2 μg/mL (100 μL/well) can bind Human GITR, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. GIR-H525a) with a linear range of 0.2-8 ng/mL (Routinely tested).
- Gene ID:AAH69319.1
- Protein Synonyms:hGITRL|TL6|AITRL|TNFSF18|GITR Ligand
- Protein/Peptide Name:GITR Ligand
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:20.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together