Order Entry
United States
ContactUsLinkComponent
Human Recombinant Frizzled-7 (from HEK293 cells)
Human Recombinant Frizzled-7 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant Frizzled-7 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Loaded Biotinylated Human Glypican 3, His,Avitag (Cat. No. GP3-H82E5) on SA Biosensor, can bind Human Frizzled-7, His Tag (Cat. No. FZ7-H52H9) with an affinity constant of 1.18 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    O75084-1
  • Protein Synonyms:
    Fz-7|FzE3|hFz7|Frizzled-7|FZD7
  • Protein/Peptide Name:
    Frizzled-7
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    18.6 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together