Human Recombinant Frizzled-2 (from HEK293 cells)
Catalog # 103790-402
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q14332-1
- Protein Synonyms:Fz-2|FZD2|hFz2|Frizzled-2|FzE2
- Protein/Peptide Name:Frizzled-2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:44.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-402
Specifications
About this item
Human Frizzled-2 / FZD2 (24-190) Protein, Fc Tag, ACROBiosystems