Human Recombinant FOLR1 (from HEK293 cells)
Catalog # 103790-392
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P15328
- Protein Synonyms:FOLR-1|FOLR|FBP
- Protein/Peptide Name:FOLR1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:27.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-392
Specifications
About this item
Human FOLR1 Protein, Strep Tag, ACROBiosystems