Human Recombinant Flt-3 Ligand (from HEK293 cells)
Catalog # 103790-386
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human Flt-3, Fc Tag (Cat. No. FL3-H5258) at 5 μg/mL (100 μL/well) can bind Human Flt-3 Ligand, His Tag (Cat. No. FLL-H5223) with a linear range of 0.1-0.6 ng/mL (QC tested).
- Gene ID:P49771-1
- Protein Synonyms:FLT3L|FLT3LG|FL|Flt3 ligand
- Protein/Peptide Name:Flt-3 Ligand
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:19.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-386
Specifications
About this item
Human Flt-3 Ligand Protein, His Tag, ACROBiosystems