Human Recombinant Flt-3 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human Flt-3 / Flk-2 Protein, Fc Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human Flt-3 Ligand, Tag Free (Cat. No. FLL-H5218) at 1 μg/mL (100 μL/well) can bind Human Flt-3, Fc Tag (Cat. No. FL3-H5258) with a linear range of 2-31 ng/mL (QC tested).
- Gene ID:P36888-1
- Protein Synonyms:FLT-3|Flk-2|STK-1|Flt-3|CD135|FLK2
- Protein/Peptide Name:Flt-3
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:84.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together