Order Entry
United States
ContactUsLinkComponent
Human Recombinant FGL1 (from HEK293 cells), Biotin
Human Recombinant FGL1 (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant FGL1 (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human LAG-3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. LA3-H525c) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGL1, Avitag,Fc Tag (Cat. No. FG1-H82F4) with a linear range of 0.156-5 μg/mL (QC tested). Loaded Biotinylated Human FGL1, Avitag,Fc Tag (Cat. No. FG1-H82F4) on SA Biosensor, can bind Human LAG-3, Fc Tag (Cat. No. LA3-H5255) with an affinity constant of 4.12 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS analysis shows that Biotinylated Human FGL1, Avitag,Fc Tag (Cat. No. FG1-H82F4) can bind to 293T cells overexpressing human LAG3. The concentration of Human FGL1 is 10 μg/mL (Routinely tested).
  • Gene ID:
    Q08830-1
  • Protein Synonyms:
    Hepassocin|HP-041|HFREP1|FGL1|LFIRE-1|HFREP-1
  • Protein/Peptide Name:
    FGL1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    62.1 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together