Human Recombinant FGL1 (from HEK293 cells), Biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human FGL1 Protein, His,Avitag™ (recommended for biopanning), ACROBiosystems
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human FGL1, His,Avitag (recommended for biopanning) (Cat. No. FG1-H82Ey) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-FGL1 antibody with a linear range of 0.01-0.625 μg/mL (QC tested). Loaded Biotinylated Human FGL1, His,Avitag (recommended for biopanning) (Cat. No. FG1-H82Ey) on SA Biosensor, can bind Human LAG-3, Fc Tag (HPLC-verified) (Cat. No. LA3-H5255) with an affinity constant of 8.12 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS assay shows that Biotinylated Human FGL1, His,Avitag (Cat. No. FG1-H82Ey) can bind to 293T cells overexpressing Human LAG3. The concentration of Human FGL1 is 10 μg/mL (Routinely tested).
- Gene ID:Q08830-1
- Protein Synonyms:Hepassocin|HP-041|HFREP1|FGL1|LFIRE-1|HFREP-1
- Protein/Peptide Name:FGL1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:37.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS with Arginine, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together