Human Recombinant Fc gamma RIIIB/ CD16b (NA1) (from HEK293 cells)
103815-522
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:50 µg
- Tag sequence:His Tag (SPR & BLI verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Immobilized Human CD16b (NA1), His Tag (SPR & BLI verified) (Cat. No. CDB-H5227) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 8.17 μM as determined in a SPR assay (Biacore T200) (QC tested). Loaded Human CD16b (NA1), His Tag (SPR & BLI verified) (Cat. No. CDB-H5227) on HIS1K Biosensor, can bind Rituximab with an affinity constant of 18 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:O75015-1(S36R,S65N,N82D,I106V)
- Protein Synonyms:Fc gamma RIIIB/ CD16b (NA1)|FCGR3B|FCGR3|FCG3B|FCG3|CD16B|IGFR3
- Protein/Peptide Name:Fc gamma RIIIB/ CD16b (NA1)
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:22.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103815-522
Human Fc gamma RIIIB / CD16b (NA1) Protein, His Tag (SPR & BLI verified), ACROBiosystems