Human Recombinant Fc gamma RIIIA/ CD16a (from HEK293 cells)
Catalog # 103815-546
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:50 µg
- Tag sequence:His Tag (SPR & BLI verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Immobilized Human CD16a (F176), His Tag (SPR & BLI verified) (Cat. No. CDA-H5220) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 0.63 μM as determined in a SPR assay (Biacore T200) (QC tested). Loaded Human CD16a (F176), His Tag (SPR & BLI verified) (Cat. No. CDA-H5220) on HIS1K Biosensor, can bind Rituximab with an affinity constant of 1.72 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Herceptin on Protein A Biosensor, can bind Human CD16a (F176), His Tag (SPR & BLI verified) (Cat. No. CDA-H5220) with an affinity constant of 0.47 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Herceptin on FAB2G Biosensor, can bind Human CD16a (F176), His Tag (SPR & BLI verified) (Cat. No. CDA-H5220) with an affinity constant of 0.51 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P08637-1
- Protein Synonyms:FCGR3A|CD16A|FCGR3|FCG3|IGFR3
- Protein/Peptide Name:Fc gamma RIIIA/ CD16a
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:23.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103815-546
Specifications
About this item
Human Fc gamma RIIIA / CD16a (F176) Protein, His Tag (SPR & BLI verified), ACROBiosystems