Human Recombinant Fc epsilon RI alpha (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human Fc epsilon RI alpha Protein, Fc Tag (BLI verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (BLI verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Loaded Human Fc epsilon RI alpha, Fc Tag (BLI verified) (Cat. No. FCA-H5259) on AHC Biosensor, can bind Immunoglobulin E, Human Plasma with an affinity constant of 0.188 nM as determined in BLI assay (ForteBio Octet Red96e) (QC Tested).
- Gene ID:P12319-1
- Protein Synonyms:FCE1A|FcERI|FCER1A
- Protein/Peptide Name:Fc epsilon RI alpha
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:47.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together