Human Recombinant Dkk-1 (from HEK293 Cells)
Catalog # 103790-316
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Tag sequence:Strep Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human Dkk-1, Strep Tag (Cat. No. DK1-H5286) at 5 μg/mL (100 μL/well) can bind Human LRP-5, Mouse IgG2a Fc Tag (Cat. No. LR5-H5254) with a linear range of 0.02-0.626 μg/mL (QC tested).
- Gene ID:O94907
- Protein Synonyms:SK|DKK1
- Protein/Peptide Name:Dkk-1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:28.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-316
Specifications
About this item
Human Dkk-1 Protein, Strep Tag, ACROBiosystems