Human Recombinant CD8 alpha & beta (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human CD8 alpha&beta (CD8A&CD8B) Heterodimer Protein, His Tag&Tag Free (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P01732-1 (CD8A) & P10966-1 (CD8B)
- Protein Synonyms:CD8A & CD8B|CD8 alpha & beta
- Protein/Peptide Name:CD8 alpha & beta
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:19.5 kDa (CD8A) & 22.1 kDa (CD8B)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together