Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD8 alpha & beta (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Human Recombinant CD8 alpha & beta (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant CD8 alpha & beta (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 ℃, aviod light
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P01732-1 (CD8A) & P10966-1 (CD8B)
  • Protein Synonyms:
    CD8A & CD8B|CD8 alpha & beta
  • Protein/Peptide Name:
    CD8 alpha & beta
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    19.5 kDa (CD8A) & 22.1 kDa (CD8B)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together