Human Recombinant CD73 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human CD73 Protein, Mouse IgG2a Fc Tag (active enzyme), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Gene ID:P21589-1
- Protein Synonyms:5'-Nucleotidase|5'-NT|NTE|NT5|NT5E|CD73
- Protein/Peptide Name:CD73
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:84.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 51 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally Trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together