Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD59 (from HEK293 Cells), Biotin
Human Recombinant CD59 (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems
Certificates
Human Recombinant CD59 (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    Precision Avi
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P13987-1
  • Protein Synonyms:
    Protectin|FLJ38134|MIRL|MEM43|1F5|MACIF|MIN3|EJ16|MSK21|EL32|MGC2354|HRF20|p18-20|MIC11|MIN1|CD59|16.3A5|MIN2|MAC-IP|G344|EJ30|FLJ92039
  • Protein/Peptide Name:
    CD59
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    12.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together