Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD3E & CD3D (from HEK293 Cells), Biotin
Human Recombinant CD3E & CD3D (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems
Certificates
Human Recombinant CD3E & CD3D (from HEK293 Cells), Biotin
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Biotinylated Human CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free (Cat. No. CDD-H82W6) captured on Biotin CAP - Series S sensor Chip can bind Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) with an affinity constant of 22.5 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free (Cat. No. CDD-C82W6) on SA Biosensor, can bind Human CD3×BCMA scFV with an affinity constant of 1.84 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P07766-1 (CD3E) & P04234-1 (CD3D)
  • Protein Synonyms:
    CD3 delta / epsilon|CD3 delta & CD3 epsilon|CD3E & CD3D
  • Protein/Peptide Name:
    CD3E & CD3D
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    18.6 kDa (CD3E) and 13.4 kDa (CD3D)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together