Cynomolgus Monkey Recombinant TRAIL R2 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Cynomolgus TRAIL R2 / DR5 / TNFRSF10B Protein, His Tag (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:A0A2K5TXK0-1
- Protein Synonyms:TRAILR2|TRAIL-R2|CD262|TRICK2|KILLER|TRICKB|ZTNFR9|TNFRSF10B|DR5
- Protein/Peptide Name:TRAIL R2
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:19.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together